DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier:  DWK Life Sciences 223742
 View more versions of this product
Catalog No. 06-406E
Explore available promotions
Description
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage
Specifications
Specifications
0.7 oz., 20 mL
0.7 oz.
1.25 in.
58 mm
32 mm
20 mL
Clear
Borosilicate Glass
2.28 in.