DWK Life Sciences Wheaton™ Serum Bottles and Vials
These bottles have exceptional wall strength and superior heat and chemical tolerances
Supplier: DWK Life Sciences 223745
Description
These bottles have exceptional wall strength and superior heat and chemical tolerances
Ideal for long- and short-term sample storage and vaccine/injectable drug containers.
- Clear borosilicate glass conforms to USP Type 1 requirements
- Amber borosilicate glass conforms to USP Type 1 requirements
- Shrink wrapped partitioned modules reduce breakage
Specifications
1.7 oz., 50 mL | |
1.7 oz. | |
1.69 in. | |
73 mm | |
43 mm |
50 mL | |
Clear | |
Borosilicate Glass | |
2.87 in. |