DWK Life Sciences Wheaton™ Serum Vials
Available as clear and amber vials
Supplier:  DWK Life Sciences 223687
 View more versions of this product
Catalog No. 06-446E
Explore available promotions
Description
Description
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Specifications
Specifications
0.51 in.
Clear
Borosilicate Glass
For lyophilization, packaging, sampling and freeze drying applications
Wheaton
Autoclavable
58 mm
Serum Vial
13 mm
30.5 mm
20 mL
5 dr.
20 mL
Crimp Cap
ASTM E438, USP Type I, Type I, Class A