DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers
Provide airtight covers
$449.59 - $883.21
Catalog Number | Mfr. No. | Color | Product Type | For Use With (Equipment) | For Use With (Application) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Product Type | For Use With (Equipment) | For Use With (Application) | Price | Quantity | ||
06-451-56
|
DWK Life Sciences
224100192 |
Gray | 2-Leg Lyophilization Stopper; Chlorobutyl Isoprene blend | Fits 13mm I.D., 20mm O.D. mouth of serum bottles and vials | Lyophilization |
Case of 1000 Each for $883.21
|
|
||
06-447E
|
DWK Life Sciences
W224100093 |
Gray | 2-Leg Lyophilization Stopper; Butyl Rubber | Fits 7mm I.D., 13mm O.D. mouth of serum bottles and vials | Lyophilization |
Case of 1000 Each for $449.59
|
|
||
06-406-11B
|
DWK Life Sciences
224100172 |
Red | Straight Plug Stopper | Fits 1.3mm I.D., 20mm O.D. mouth of serum bottles and vials | Not for lyophilization |
Case of 1000 Each for $621.91
|
|
||
Description
Provide airtight covers
- Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
- Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
Specifications
Autoclavable |