DWK Life Sciences Wheaton™ V Vial™ with Solid-Top Screw Cap, Clear
Conical interior bottoms are designed to provide total downward drainage and maximum retrieval of contents by syringe
Supplier:  DWK Life Sciences W986259NG
 View more versions of this product
Catalog No. 22-281410
Explore available promotions
Includes: 14B rubber-lined black phenolic screw caps faced with PTFE
Description
Description
Clear vials are manufactured from Wheaton-33™ low extractable borosilicate glass
Specifications
Specifications
PTFE/14B Rubber
20 mm
5.0 mL
No
Screw
Phenolic
65 mm
20-400
Clear
Borosilicate Glass (Low Extractable)
Ideal for small scale reactions, centrifugation, storage, packaging and shipping
Conical V
1.4 dr.
Screw Cap
ASTM E438, USP Type I, Type I, Class A