missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nasco Whirl-Pak™ Carrying Racks
For transporting and using sample-filled bags in laboratories
Supplier: Nasco B00677WA
Description
For transporting and using sample-filled bags in laboratories
- Autoclavable
- 10 and 12 gauge steel
- Electrostatically applied powder coat finish
- Max. dry temperature 300°F and wet 500°F
- Easy sample transport and storage
Specifications
Autoclavable | |
Coated Wire | |
1.8 x 16.5 x 7.6 cm (12.5 x 6.5 x 3 in.) | |
16.5 cm | |
7.6 cm |
Bags (21) | |
21 | |
12.5 in. | |
6.5 in. | |
3 in. |