Nasco Whirl-Pak™ Thio-Bag™ PE Bags for Chlorinated Water Sampling
Ideal for sampling potable water, swimming pool water, effluents and industrial wastes
Supplier:  Nasco B01254
 View more versions of this product
Catalog No. 01-812-8
Explore available promotions
Description
Description
Ideal for sampling potable water, swimming pool water, effluents and industrial wastes
Unbreakable, sterile bags to collect and dechlorinate water and wastewater samples for bacteriological examination in compliance with EPA and APHA requirements.
  • Economical and easy to dispose
  • 4 oz. (118 mL) bag contains one nontoxic, non-nutritive pill impregnated with 10 mg sodium thiosulfate and has 100 mL fill-line
  • 18 oz. (532 mL) bag contains one 25 mg sodium thiosulfate pill and has 300 mL fill-line
Specifications
Specifications
Standard Whirl-Pak Thio-Bags
532 mL
Contains one 25 mg sodium thiosulfate pill.
Sterile
100/Pk.
2.50 mil
9 in.
4.5 in.
18 oz.
Sampling chlorinated water
Polyethylene
0.064 mm
82°C
23 x 11.5 cm (9 x 4.5 in.)
23 cm
11.5 cm