Nasco Whirl-Pak™ Write-On Stand-Up Thio Sample Bag
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
Supplier:  Nasco B01403
 View more versions of this product
Catalog No. 01-812-123
Explore available promotions
Description
Description
Bag stands up completely by itself and contains 10 mg of active sodium thiosulfate to neutralize chlorine in water samples.
  • A special gusset on the bottom of the bag flattens out when water is put in the bag, allowing it to stand up completely by itself.
  • No rack or other holder is needed to keep the bag upright.
  • Forget washing and sterilizing glass bottles and disposing of expensive vials.
  • Sterile and unbreakable
Specifications
Specifications
Stand-up Bag
300 mL
25 bags/pouch, 100 bags/box
23 x 11.5 cm (9 x 4.5 in.)
23 cm
11.5 cm
10 oz.
0.076 mm
3 mil
9 in.
4.5 in.