missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ White 96-Well Immuno Plates
Conduct quantitative and qualitative immunoassays like ELISA and binding assays with these white, 96-well immuno plates.
$675.30 - $1815.50
Specifications
Format | Standard |
---|---|
Array | 8 x 12 |
Color | White |
Product Type | Microplate |
No. of Wells | 96 |
Catalog Number | Mfr. No. | Surface Treatment | Well Shape | Volume (Metric) Working | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Surface Treatment | Well Shape | Volume (Metric) Working | Price | Quantity | |||
12-566-05
|
Thermo Scientific™
436110 |
MaxiSorp | Round, Flat Bottom (F-well) | 350 μL |
Case of 80 Each for $1,538.63
|
|
|||
12-566-06
|
Thermo Scientific™
436111 |
PolySorp | Round, Flat Bottom (F-well) | 350 μL |
Case of 80 Each for $1,815.50
|
|
|||
14-245-196P
|
Thermo Scientific™
7571 |
Microlite 1+ | Round, Flat Bottom (F-well) | 280 μL |
Case of 50 Each for $675.30
|
|
|||
14-245-196Q
|
Thermo Scientific™
7572 |
Microlite 2+ | Round, Flat Bottom (F-well) | 280 μL |
Case of 50 Each for $822.43
|
|
|||
14-245-197B
|
Thermo Scientific™
7705 |
Microfluor 1 | Round, Flat Bottom (F-well) | 280 μL |
Case of 50 Each for $692.18
|
|
|||
14-245-178
|
Thermo Scientific™
7905 |
Microfluor 2 | Round, Flat Bottom (F-well) | 280 μL |
Case of 50 Each for $827.25
|
|
|||
21-377-273
|
Thermo Scientific™
9502887 |
Universal Binding | Round, Flat Bottom (F-well) | 330 μL |
Case of 50 Each for $1,191.06
|
|
|||
Description
Conduct quantitative and qualitative immunoassays, like ELISA and binding assays, with Thermo Scientific™ White 96-Well Immuno Plates. These plates, which are compatible with common instruments, deliver optimal results, lot-to-lot reliability and well-to-well reproducibility.
Features
- Plate has SBS footprint — fits standard equipment
- Available with flat- or round-bottom wells
- Choice of three surface types for optimal binding
- Certified binding homogeneity/reproducibility
Choice of Surface:
- Thermo Scientific PolySorp, Microfluor 1, Microlite 1+ and Universal Binding (UB) for adsorption of hydrophobic molecules
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
- Thermo Scientific Microfluor 2 and Microlite 2+ are slightly hydrophilic and bind a diverse range of biomolecules
White:
- Gives maximum reflection
- Minimum autofluorescence and autoluminescence
Compatible with:
Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane
Ordering Information:
Availability may vary by country.
Specifications
Standard | |
White | |
96 |
8 x 12 | |
Microplate |