missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wells Lamont™ White/Blue Performance Crew Socks
White or blue socks, constructed of polyester/nylon/spandex, are ideal for cleanrooms.
$321.07 - $332.59
Specifications
For Use With (Application) | Cleanroom Use |
---|---|
Material | Polyester, Nylon, Spandex |
Color | White, Blue |
Description
- Men's Performance Work Socks provide support and protection for your feet while you work.
- The mesh upper foot is designed for ventilation and moisture evaporation, keeping your feet dry throughout the day.
- Socks feature reinforced heel and toe as well as an Achilles heel pad for added protection and comfort.
- The socks non-binding top do not slip down and provide all-day gentle comfort.
- Added compression at the arch provides added support and stability.
Specifications
Cleanroom Use | |
Polyester, Nylon, Spandex | |
White, Blue |