Thermo Scientific™ White Disposable Shaker/Incubator Trays
Support the Thermo Scientific™ Oxoid™ Signal™ Blood Culture System during shaking and incubation using these disposable shaker/incubator trays.
Supplier:  Thermo Scientific™ BC0104A
Catalog No. OXBC104A
Explore available promotions
Description
Description
Support the Thermo Scientific™ Oxoid™ Signal™ Blood Culture System during shaking and incubation using Thermo Scientific™ Oxoid™ Signal™ Disposable Shaker/Incubator Trays. The Signal Blood Culture System is a unique one bottle manual media system designed to detect aerobic, anaerobic and microaerophilic organisms.
Signal Blood Culture System (BC0100M) comes complete with blood culture bottles filled with the blood culture medium and growth indicator devices. Incubator Trays can be ordered separately, where required.
A single white Disposable Shaker/ Incubator Tray can hold up to 10 bottles.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
Disposable Shaker/Incubator Trays
5/Pk.