Fisherbrand™ White Polypropylene Caps
White polypropylene screw caps provides clear surface for writing on cap
 View more versions of this product
Catalog No. FB02912069
Explore available promotions
Description
Description
  • Foamed polyethylene liner provides tight seal and chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
  • PTFE faced foamed polyethylene liner offers chemical resistance excellent for environmental samples, pharmaceuticals and diagnostic reagents
  • Poly-vinyl liner provides an excellent moisture barrier good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Specifications
Specifications
White
White
No
PTFE/Polyethylene Foam
100
24-400
Lined Cap
Polypropylene