missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Supelco™ White/Tan PTFE/Silicone Septa

May be used in headspace solid-phase microextraction (SPME) (lyophilized aqueous extract)

Supplier:  MilliporeSigma™ Supelco™ 27155

 View more versions of this product

Catalog No. 111011144


Explore available promotions

Description

Description

  • A gas tight and inert nature septa, which is used for high temperature applications during pretreatment and sample oxidation.
Specifications

Specifications

13 mm
PTFE/Silicone
Temperature limit 120°C
7 mL Vial
Tan, White
100
0.06 in.
Videos
SDS
Documents

Documents

Promotions

Promotions