United Scientific™ Whiteboard Optics Set DFS Item
Set of equipment for ray optics and color experiments using magnetic-backed components.
Supplier:  United Scientific™ WBOPS1A
Catalog No. S15791
Explore available promotions
Description
Description
  • Collection of educational assets designed for lessons on optics. All optical elements are 23 mm thick, with built-in magnets and white-painted bottom surfaces.
  • Optics Set Contains:
    • 1 double convex lens, 75 mm wide
    • 1 double concave lens, 75 mm wide
    • 1 semicircular lens, 75 mm wide
    • 1 equilateral prism, 60 mm
    • 1 right-angle prism, 50 mm (2 in.)
    • 1 30°/60°/90° prism, 38 x 62 x 73 mm
    • 1 rectangular block, 75 x 50 mm
    • 1 rectangular trough, 61 mL, 75 x 50 mm (3 x 2 in.)
    • 1 plane mirror, 10 cm
    • 1 adjustable concave/convex mirror, 135 mm
    • Whiteboard has a usable surface of 23 x 15.5 in. with swiveling feet on two sides
    • Allow for setup in wide or tall format
  • Available as a full set, optical accessories only, or whiteboard only
Specifications
Specifications
Acrylic
Optical Parts Only
Optics Set
Various
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm