CoorsTek™ Wide-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
$57.63 - $57.63
Specifications
Material Porcelain
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
07-955F
View Documents
CoorsTek™
CRS66137
50 mL
Each for $57.63
Case of 24 Each for $1,383.12
Description
Description
  • Popular, low-form crucibles
  • Glazed inside and out except for outside bottom surface and rim
  • Covers sold separately
Specifications
Specifications
Porcelain
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'