missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Wide Mouth Glass Reagent Bottle with Stopper
Wide Mouth Glass Reagent Bottle with Stopper is made from soda glass and features a ground glass stopper and a wide mouth design.
Supplier: Eisco™ CH0156FW
Description
- Glass reagent bottle with ground glass stopper
- Made from soda glass
- Wide mouth design for easy filling and a wide pour lip
- Glass stopper seals the bottle to prevent evaporation
Specifications
1000 mL | |
34 oz. | |
4.1 in. | |
Soda Lime Glass | |
1 Ea. | |
10.5 cm | |
Autoclavable | |
No | |
Cylindrical | |
Reagent Bottle | |
Glass | |
10.5 cm |
1000 mL | |
Clear | |
Holding liquids | |
10.5 cm | |
Non-sterile | |
Yes | |
Reusable | |
Wide | |
21.6 cm | |
Blank | |
5 cm | |
Stopper |