missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Wilmad™ Multi-tube Jet Solvent NMR/EPR Tube Washer/Dryer

Accommodate up to 5 tubes (3, 4 and 5 mm OD) at once. Bel-Art SP Scienceware™ Wilmad™ Multi-tube Jet Solvent NMR/EPR Tube Washer/Dryer includes calibrated length marks for 4 in., 7 in., 8 in. and 9 in. tubes.

$1328.00 - $1328.00

Specifications

Product Type 5-tube Washer
Diameter (Metric) Outer For tubes 3 to 5mm O.D.
Includes Washer body, solvent cup, and solvent transfer tubing.
View More Specs

Products 1
Catalog Number Mfr. No. Model Description Price Quantity  
Catalog Number Mfr. No. Model Description Price Quantity  
16-800-167
View Documents
Bel-Art™
660000067
WG-1209-J1 Joint type: 24/40 Taper Joint
Each for $1,328.00
 
Description

Description

  • Conveniently provides hands-free operation throughout the washing and drying cycle
  • PTFE solvent cup and PTFE coated transfer tubing eliminate scratches and makes this unit fully resistant to common organic solvents
  • Flanged reservoir connection avoids joint freeze while a greaseless joint between the solvent cup and glass reservoir remains contamination free
  • Disassembles completely without tools for easy cleaning
  • Requires filter flask (sold separately; 1000ml recommended)
Specifications

Specifications

5-tube Washer
For tubes 3 to 5mm O.D.
Washer body, solvent cup, and solvent transfer tubing.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'