Learn More
Eisco™ Wimshurst Machine
The Wimshurst Machine generates electrical charges.
Supplier:  Eisco™ PH0848C
Catalog No. S05839
Only null left
Explore available promotions
Description
Description
  • Heavy, high resistance 11.8 in. (300 mm) disks with sectors of aluminum strips are belt-driven in opposing directions.
  • Leyden jars of glass and aluminum foil capture charges.
  • Disks supported by two rigid uprights.
  • Mounted on polished wooden base.
Specifications
Specifications
7 to Undergraduate
Transparent plates with radial aluminum segments driven in opposite directions by hand wheel and belt, with 2 leyden jars, mounted on moulded abs base, instructions
5.39 kg
1 Class
Ideal classroom demonstration device for electrical and physics experiments
11.8 lb.
19.68 in.
Wimshurst Machine
Product Suggestions
Product Suggestions