missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wind-Up Measuring Tapes
Choose from three lengths
$34.31 - $34.31
Specifications
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
S94239
|
Customer Channels Group (Ccg)
LER0365 |
Each for $34.31
|
|
||||||
Description
- Choose from 33 ft. (10m), 100 ft. (30m), or 165 ft. (50m) lengths
- Made of fiberglass
- 100 ft. and 165 ft. models have removable handle for easy control
- Marked in cm and mm on one side and inches on reverse
- Rolls into sturdy case
Specifications
0.0 |