Windup Meter Tapes
Durable tapes won't tear, break, rust or stretch.
$13.18 - $33.21
Specifications
Products 3
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S94309
View Documents
Customer Channels Group (Ccg)
45200
Each for $13.18
S94310
View Documents
Customer Channels Group (Ccg)
45480
Each for $25.97
S94311
View Documents
Customer Channels Group (Ccg)
45500
Each for $33.21
Description
Description
  • High-quality fiberglass in ABS plastic windup case
  • Calibrated on one side in centimeters and on the other in inches
  • Great for outdoor activities, track and field, rough terrain, mud and water
Specifications
Specifications
0.0
SDS
missing translation for 'documents'
missing translation for 'documents'