Poltex Wipe Canister and Glove Box Holder
A break-resistant PETG wipe canister and glove box holder that holds two boxes of gloves and a wipe tub.
$363.00 - $459.79
Specifications
Product Type Wipe Dispenser
Material PETG
Capacity 2 Boxes
View More Specs
Products 3
Catalog Number Mfr. No. Mounting Price Quantity  
Catalog Number Mfr. No. Mounting Price Quantity  
22000731
View Documents
Poltex
GBSTUFM
Magnetic Mount
Each for $459.79
22000732
View Documents
Poltex
GBSTUFT
VHB Tape
Each for $435.62
22000733
View Documents
Poltex
GBSTUFW
Wall Mount
Each for $363.00
Description
Description
  • Rubber bumpons on base for non-slip placement on countertop.
  • Choose from wall mount/countertop placement (screws/anchors included), magnet mount (powerful magnets included), or tape mount (VHB tape included).
Specifications
Specifications
Wipe Dispenser
PETG
2 Boxes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'