Brady™ Wire and Cable Identification BBP33 Labels: One per Row
Wire and Cable Identification BBP33 Labels: One per Row are made of self-laminating vinyl or nylon cloth, which are excellent materials for wire and cable identification.
Supplier:  Brady™ B33118427
 View more versions of this product
Catalog No. 19-140-681
Explore available promotions
Description
Description
  • The adhesive and cloth backing material (B-427) offers excellent holding power, yet allow for clean removal and repositioning. Sticks to any clean, dry surface and is specially formulated topcoat for excellent print quality.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
Specifications
Specifications
RoHS, UL
White, Translucent
Vinyl
Wire and cable label
1250/Pk.
-40°C to +70°C
0.004 in.
1 in.
Acrylic, Permanent
Translucent
Identification Label
BBP™33 Printer
-40 to 158°F
12/11 lb./in.
1.5 in.
2.5 cm