missing translation for 'onlineSavingsMsg'
Learn More

WNT3A Polyclonal Antibody, PeproTech®, Invitrogen™ DFS Item

Rabbit Polyclonal Antibody

$337.00 - $2935.00

Specifications

Antigen WNT3A
Concentration 0.1-1.0 mg/mL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
500P25150UG DFS Item
View Documents
PeproTech
500P25150UG
50 μg
Each for $337.00
Only null left
 
500P251100 DFS Item
View Documents
PeproTech
500P251100UG
100 μg
Each for $432.00
Only null left
 
500P2511MG DFS Item
View Documents
PeproTech
500P2511MG
1 mg
Each for $2,935.00
Only null left
 
Description

Description

Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-3a. Anti-Human Wnt-3a-specific antibody was purified by affinity chromatography employing an immobilized Human Wnt-3a matrix. Sandwich ELISA: To detect Human Wnt-3a by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Wnt-3a (500-P251Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Wnt-3a. Western Blot: To detect Human Wnt-3a by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Wnt-3a is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications

Specifications

WNT3A
ELISA, Western Blot
Unconjugated
Rabbit
Human
P56704
89780
E.coli-derived Recombinant Human Wnt-3a.
Antigen affinity chromatography
WNT3A
0.1-1.0 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative
MGC119418; MGC119419; MGC119420; Protein Wnt-3a; vestigial tail; vt; wingless-related MMTV integration site 3A; wingless-type MMTV integration site family member 3A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3A; Wnt 3a; Wnt family member 3A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3a; WNT3A protein; wnt3l; Zwnt[a]
WNT3A
Primary
-20°C
Videos
SDS
Documents

Documents