missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Wood Wall Cabinet, 42 in. Wide
Fisherbrand™ Wood Wall Cabinets bring quality, warmth and durability to your laboratory.
Description
- Made from hand selected premium rotary cut maple or rift cut oak veneers
- Robotically applied environmentally friendly finishes are chemically resistant.
- Wall Cabinets come with two adjustable shelves.
- Framed Glass Doors will have tempered safety glass and have solid wood rails and stiles.
Compliance
SEFA 8
Specifications
2 years | |
Cabinet | |
79.4 cm | |
42 in. | |
30.5 cm | |
2 | |
2 | |
Maple Veneer |
Left and Right | |
12 in. | |
31.25 in. | |
Framed Glass Door | |
106.7 cm | |
Wall | |
Amber Stain |