missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Wood Wall Cabinet, 42 in. Wide

Fisherbrand™ Wood Wall Cabinets bring quality, warmth and durability to your laboratory.

 View more versions of this product

Catalog No. FB4792HM4


Explore available promotions

Description

Description

  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Wall Cabinets come with two adjustable shelves.
  • Framed Glass Doors will have tempered safety glass and have solid wood rails and stiles.

Compliance

SEFA 8

Specifications

Specifications

2 years
Cabinet
79.4 cm
42 in.
30.5 cm
2
2
Maple Veneer
Left and Right
12 in.
31.25 in.
Framed Glass Door
106.7 cm
Wall
Auburn Stain
Videos
SDS
Documents

Documents

Promotions

Promotions