Bel-Art™ Write-On™ Paper Label Tapes
Use a pencil, ball point pen or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Bel Art™ SP Scienceware™ Write-On™ Paper Label Tapes resist moisture and most solvents and are great for use in low-temperature applications.
$51.27 - $305.88
Specifications
Autoclavable Autoclavable
Adhesive Type Rubber, Resin
Material Paper
For Use With (Application) For use in low temperature applications
View More Specs
Products 15
Catalog Number Mfr. No. Diameter (English) Length (English) Width (English) Color Price Quantity  
Catalog Number Mfr. No. Diameter (English) Length (English) Width (English) Color Price Quantity  
03-411-607
View Documents Promotion Details
Bel-Art™
F134820075
1 in. 15 yd. 0.75 in. Green
Pack of 4 Each for $84.97
Case of 4 Pack for $339.88
03-411-601
View Documents Promotion Details
Bel-Art™
F134800075
1 in. 15 yd. 0.75 in. White
Pack of 4 Each for $61.65
Case of 4 Pack for $246.60
03-411-619
View Documents Promotion Details
Bel-Art™
F134860075
3 in. 40 yd. 0.75 in. Green
Pack of 4 Each for $202.09
03-411-609
View Documents Promotion Details
Bel-Art™
F134830050
3 in. 40 yd. 0.5 in. White
Pack of 6 Each for $217.56
03-411-628
View Documents Promotion Details
Bel-Art™
F134900075
3 in. 60 yd. 0.75 in. White
Pack of 4 Each for $223.44
03-411-610
View Documents Promotion Details
Bel-Art™
F134830075
3 in. 40 yd. 0.75 in. White
Pack of 4 Each for $192.50
03-411-604
View Documents Promotion Details
Bel-Art™
F134810075
1 in. 15 yd. 0.75 in. Yellow
Pack of 4 Each for $76.21
Case of 4 Pack for $304.84
03-411-602
View Documents Promotion Details
Bel-Art™
F134800100
1 in. 15 yd. 1 in. White
Pack of 3 Each for $60.15
Case of 4 Pack for $240.60
03-411-608
View Documents Promotion Details
Bel-Art™
F134820100
1 in. 15 yd. 1 in. Green
Pack of 3 Each for $78.90
Case of 4 Pack for $315.60
03-411-611
View Documents Promotion Details
Bel-Art™
F134830100
3 in. 40 yd. 1 in. White
Pack of 3 Each for $169.91
03411614
View Documents Promotion Details
Bel-Art™
F134840100
3 in. 40 yd. 1 in. Red
Pack of 3 Each for $170.24
03-411-606
View Documents Promotion Details
Bel-Art™
F134820050
1 in. 15 yd. 0.5 in. Green
Pack of 6 Each for $51.27
Case of 4 Pack for $205.08
03-411-620
View Documents Promotion Details
Bel-Art™
F134860100
3 in. 40 yd. 1 in. Green
Pack of 3 Each for $189.82
11-865-109
View Documents Promotion Details
Bel-Art™
134620300
1 in. 15 yd. 0.75 in. White, Yellow, Green
Pack of 3 Each for $68.91
11-865-110
View Documents Promotion Details
Bel-Art™
F134630600
3 in. 15 yd. 0.75 in. Assorted
Pack of 6 Each for $305.88
Description
Description
Use pencil, ball-point pen or solvent ink marker to create labels that remain legible under the most demanding laboratory conditions. Bel Art™ SP Scienceware™ Write-On™ Paper Label Tapes resist moisture and most solvents and are great for use in low temperature applications.
  • Adheres to any clean surface, including PTFE fluoropolymer resin coated materials
  • Peels off without leaving a sticky residue
  • Pencil, ball-point pen, or solvent ink marker remain legible under the most demanding laboratory conditions.
  • Colored paper with rubber resin adhesive resists moisture and most solvents
  • Withstands a maximum temperature of 157°C (315°F) for 30 minutes or 80°C (176°F) for 24 hours; can be used down to -80°C (-112°F).
  • Withstands a steam autoclaving cycle at 121°C (250°F) and gamma irradiation between 13.0 and 45.0kGy
Specifications
Specifications
Autoclavable
Rubber, Resin
Paper
For use in low temperature applications