missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ Write On Right-To-Know Laboratory Labels, Paper

RTK labels use color coding system in conjunction with written text to identify chemical, physical and health hazards. Brady Write On Right-To-Know Laboratory Labels allow user to customize message to suit needs.

Supplier:  Brady™ 58268

Catalog No. 19-107-227


Explore available promotions

Description

Description

  • To identify the chemical and its physical and health hazards
  • Allows to customize message to suit needs
  • Quantity: 500 per roll
Specifications

Specifications

HMIG Color Bar
Gloss
Write On Right-To-Know Laboratory Label
500/Roll
1.25 x 2 in. (W x H)
Black, Blue, Red, Yellow, White
Paper (B-235)
Legend: HEALTH HAZARD FLAMMABILITY HAZARD INSTABILITY HAZARD PERSONAL PROTECTION
Rectangle
31.7 x 50.8 mm (W x H)
SDS
Documents

Documents

Promotions

Promotions