Mettler Toledo™ WXTS University Edition Microbalance
The Mettler Toledo™ WXTS3DU microbalance is a great choice for academic research and testing. The Swiss engineered balance offers a rugged and durable design with simple operation. With a capacity up to 1.2 g / 3.2 g and readability from 0.001 mg / 0.01 mg this balance is equipped to deliver consistent and fast results every time.
Supplier:  Mettler Toledo™ 30376748
Catalog No. 01-914-470
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The WXTS3DU is a great solution for Academic Research and Testing. The successor to the XS3DU Microbalance is a durable solution that will offer reliable results for many years to come.
  • Swiss Engineered and Manufactured
  • Intuitive Touchscreen Operation
  • Robust Aluminum Housing 
  • Easy and Efficient Cleaning
  • Durable Magnetic Force Restoration Weighing Sensor
  • FACT - Fully Automatic Calibration Technology Internal Adjustment
  • Two AUX Ports for External Switches
  • Integrated RS232 Interface
  • Slot for optional RS, Ethernet or Bluetooth
  • Centrally Placed Level Bubble
  • Data Transfer Key to PC Host or System
Specifications
Specifications
FACT Internal Adjustment
0.001 mg, 0.01 mg (fine / full range)
1.4 mg (@ USP) (measured at 0.2g); 0.14 mg (@ U=1%, k=2) (measured at 0.2g)
0.0008 mg (measured at 0.2g)
Micro Balance
1.2 g, 3.2 g (fine/ full range)
Aluminum Housing
WXTS
<8 sec.
32 mm