Kimberly-Clark Professional™ WypAll™ X70 Wipers
Ideal rag replacements
$98.01 - $200.30
Specifications
For Use With (Application) | Ideal for all manufacturing and maintenance and repair |
---|
Catalog Number | Mfr. No. | Color | Length (English) | Width (English) | Dimensions (L x W) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Length (English) | Width (English) | Dimensions (L x W) | Price | Quantity | |||
17-986-93
|
Kimberly-Clark Professional™
41100 |
White | 16.6 in. | 16.6 in. | 16.6 x 14.9 in. (42.16 x 37.84 cm) |
Case of 300 Each for $98.01
|
|
|||
19-014-872
|
Kimberly-Clark Professional™
41412 |
Blue | 16.8 in. | 16.8 in. | 16.8 x 9.1 in. (42.67 x 23.11 cm) |
Case of 1000 Each for $198.50
|
|
|||
06-666-67
|
Kimberly-Clark Professional™
41600 |
White | 13.4 in. | 13.4 in. | 13.4 x 12.5 in. (34 x 31.75 cm) |
Case of 870 Each for $135.57
|
|
|||
19-014-866
|
Kimberly-Clark Professional™
41702 |
White | 13.4 in. | 13.4 in. | 13.4 x 9.8 in. (34 x 24.89 cm) |
Case of 825 Each for $137.03
|
|
|||
15-235-81
|
Kimberly-Clark Professional™
41200 |
White | 13 in. | 14.4 in. | 13 x 12.5 in. (33 x 31.75 cm) |
Case of 912 Each for $149.42
|
|
|||
19-137-201
|
Kimberly-Clark Professional™
06354 |
White/Red | 23.5 in. | 12.5 in. | 23.5 x 12.5 in. (59.69 x 31.75 cm) |
Case of 300 Each for $101.70
|
|
|||
19-812-925
|
Kimberly-Clark Professional™
41455 |
White | 16.8 in. | 16.8 in. | 16.8 x 9.1 in. (42.67 x 23.11 cm) |
Case of 1000 Each for $200.30
|
|
|||
Description
- Wipers combine the best qualities of disposable wipers and scrap rags
- Super absorbent industrial cleaning wipers can be used almost anywhere to clean oil, solvents — virtually any industrial spill
- Made from fast-absorbing Hydroknit™ material, an extra-strong nonwoven fabric blend that won't tear or fall apart even during heavy-duty cleaning
- Textured design won't scratch delicate surfaces
Order Info
Available in quarterfold, flat, rolls and two dispensers in blue, white or red (food service).
Specifications
Ideal for all manufacturing and maintenance and repair |