missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xanthan Gum, TCI America™

Supplier:  TCI America X0048500G

 View more versions of this product

Catalog No. X0048500G


Explore available promotions

Description

Description

Specifications

Specifications

Xanthan Gum
C8H14Cl2N2O2
500 g
VXYWXJXCQSDNHX-UHFFFAOYSA-N
2-(2,4-diaminophenoxy)ethanol;dihydrochloride
47932
11138-66-2
MFCD00131256
2-2,4-diaminophenoxy ethanol dihydrochloride, 2,4-diaminophenoxyethanol dihydrochloride, 2,4-diaminophenoxyethanol hcl, unii-z5y843j6kq, ethanol, 2-2,4-diaminophenoxy-, dihydrochloride, 2,4-diaminophenoxyethanol 2hcl, 2-2,4-diaminophenoxy ethanol hydrochloride, ethanol, 2-2,4-diaminophenoxy-, hydrochloride 1:2, 4-2-hydroxyethoxy-1,3-phenylenediamine dihydrochloride, 2,4-diaminophenoxyethanol hydrochloride
C1=CC(=C(C=C1N)N)OCCO.Cl.Cl
241.112
Crystalline Powder

Chemical Identifiers

11138-66-2
241.112
VXYWXJXCQSDNHX-UHFFFAOYSA-N
47932
C1=CC(=C(C=C1N)N)OCCO.Cl.Cl
C8H14Cl2N2O2
MFCD00131256
2-2,4-diaminophenoxy ethanol dihydrochloride, 2,4-diaminophenoxyethanol dihydrochloride, 2,4-diaminophenoxyethanol hcl, unii-z5y843j6kq, ethanol, 2-2,4-diaminophenoxy-, dihydrochloride, 2,4-diaminophenoxyethanol 2hcl, 2-2,4-diaminophenoxy ethanol hydrochloride, ethanol, 2-2,4-diaminophenoxy-, hydrochloride 1:2, 4-2-hydroxyethoxy-1,3-phenylenediamine dihydrochloride, 2,4-diaminophenoxyethanol hydrochloride
2-(2,4-diaminophenoxy)ethanol;dihydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions