missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Xanthane Hydride 95.0+%, TCI America™
Supplier: TCI America X000125G
Specifications
Xanthane Hydride | |
C2H2N2S3 | |
25 g | |
YWZHEXZIISFIDA-UHFFFAOYSA-N | |
5-amino-1,2,4-dithiazole-3-thione | |
2735329 | |
≥95.0% (HPLC) |
6846-35-1 | |
MFCD00051660 | |
xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol | |
C1(=NC(=S)SS1)N | |
150.232 | |
150.23 | |
Crystalline Powder |
Chemical Identifiers
6846-35-1 | |
150.232 | |
YWZHEXZIISFIDA-UHFFFAOYSA-N | |
2735329 | |
C1(=NC(=S)SS1)N |
C2H2N2S3 | |
MFCD00051660 | |
xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol | |
5-amino-1,2,4-dithiazole-3-thione |
Safety and Handling
RTECSNumber : JO6500000
TSCA : No