missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xanthane Hydride 95.0+%, TCI America™

Supplier:  TCI America X0001500G

 View more versions of this product

Catalog No. X0001500G


Explore available promotions

Description

Description

Specifications

Specifications

Xanthane Hydride
C2H2N2S3
500 g
YWZHEXZIISFIDA-UHFFFAOYSA-N
5-amino-1,2,4-dithiazole-3-thione
2735329
≥95.0% (HPLC)
6846-35-1
MFCD00051660
xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol
C1(=NC(=S)SS1)N
150.232
150.23
Crystalline Powder

Chemical Identifiers

6846-35-1
150.232
YWZHEXZIISFIDA-UHFFFAOYSA-N
2735329
C1(=NC(=S)SS1)N
C2H2N2S3
MFCD00051660
xanthane hydride, 3-amino-1,2,4-dithiazole-5-thione, 5-imino-1,2,4-dithiazolidine-3-thione, xanthanohydrogen, isoperthiocyanic acid, 5-amino-3h-1,2,4-dithiazole-3-thione, xanthahydrogen, 1,2,4-dithiazolidine-3-thione, 5-imino, 1,2,4-dithiazole-3-thione, 5-amino, 5-imino-5h-1,2,4 dithiazole-3-thiol
5-amino-1,2,4-dithiazole-3-thione
Videos
Safety and Handling

Safety and Handling

RTECSNumber : JO6500000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions