missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xanthine, 99%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals A1107722

 View more versions of this product

Catalog No. AAA1107722


Explore available promotions

Description

Description

Xanthine and xanthine oxidase system can be used to produce superoxide radicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xanthine and xanthine oxidase system can be used to produce superoxide radicals.

Solubility
Soluble in water(0.067g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

Xanthine
>300°C
C5H4N4O2
100 g
14,10059
Soluble in water(0.067g/L).
C1=NC2=C(N1)C(=O)NC(=O)N2
152.113
CHEBI:17712
99%
69-89-6
Odorless
MFCD00078453
8733
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
LRFVTYWOQMYALW-UHFFFAOYSA-N
3,7-dihydropurine-2,6-dione
1188
152.11

Chemical Identifiers

69-89-6
152.113
LRFVTYWOQMYALW-UHFFFAOYSA-N
1188
3,7-dihydropurine-2,6-dione
C5H4N4O2
MFCD00078453
xanthine, 2,6-dihydroxypurine, xanthin, 2,6-dioxopurine, pseudoxanthine, 1h-purine-2,6 3h,7h-dione, isoxanthine, xanthic oxide, 1h-purine-2,6-diol, 9h-purine-2,6-diol
CHEBI:17712
C1=NC2=C(N1)C(=O)NC(=O)N2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-718-6

RTECSNumber : ZD7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only