missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Xylene cyanol FF, ∽75%, Thermo Scientific Chemicals
$113.34 - $113.34
Chemical Identifiers
CAS | 2650-17-1 |
---|---|
Molecular Formula | C25H27N2NaO6S2 |
Molecular Weight (g/mol) | 538.61 |
MDL Number | MFCD00019481 |
InChI Key | VVLFAAMTGMGYBS-KRQUPCAFSA-M |
Synonym | Acid blue 147 monosidium salt, C.I. 42135 |
IUPAC Name | sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate |
SMILES | [Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC440700100
|
Thermo Scientific Chemicals
440700100 |
10 g | Glass bottle |
Each for $113.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2650-17-1 | |
538.61 | |
VVLFAAMTGMGYBS-KRQUPCAFSA-M | |
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate |
C25H27N2NaO6S2 | |
MFCD00019481 | |
Acid blue 147 monosidium salt, C.I. 42135 | |
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O |
Specifications
2650-17-1 | |
MFCD00019481 | |
VVLFAAMTGMGYBS-KRQUPCAFSA-M | |
sodium 4-{[4-(ethylamino)-3-methylphenyl][(1Z,4Z)-4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl}-3-sulfobenzene-1-sulfonate | |
538.61 | |
Blue/Gray to Green | |
Powder | |
∽75% |
C25H27N2NaO6S2 | |
Acid blue 147 monosidium salt, C.I. 42135 | |
[Na+].CCNC1=C(C)C=C(C=C1)C(=C1/C=C\C(=N\CC)\C(C)=C1)\C1=C(C=C(C=C1)S([O-])(=O)=O)S(O)(=O)=O | |
538.61 | |
Glass bottle | |
10 g | |
Xylene cyanol FF |
RUO – Research Use Only