missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Xylene, Fisherbrand™, HistoPrep™

Supplier: Epredia HC7001GAL
Specifications
Liquid | |
-34°C | |
1330-20-7 | |
8.29mmHg at 25°C | |
1 gal. | |
Dimethylbenzene, xylol, methyl toluene, h-val-nh2 . hbr, c5h12n2o.hbr, l-valinamide hydrobromide, s-2-amino-3-methylbutanamide hydrobromide, h-val-nh2 inverted exclamation mark currency hbr, 2s-2-amino-3-methylbutanamide hydrobromide | |
CC1=CC(C)=CC=C1 | |
106.17 | |
106.17 |
Xylene | |
136°C | |
Poly Bottle | |
Clear | |
C8H10 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
1,3-xylene | |
57364729 |
Chemical Identifiers
1330-20-7 | |
106.17 | |
Dimethylbenzene, xylol, methyl toluene, h-val-nh2 . hbr, c5h12n2o.hbr, l-valinamide hydrobromide, s-2-amino-3-methylbutanamide hydrobromide, h-val-nh2 inverted exclamation mark currency hbr, 2s-2-amino-3-methylbutanamide hydrobromide | |
1,3-xylene |
C8H10 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
57364729 | |
CC1=CC(C)=CC=C1 |