Learn More
Xylenes, 96%, pure, mixed isomers with ethylbenzene, Thermo Scientific Chemicals
$128.97 - $203.72
Chemical Identifiers
CAS | 1330-20-7 |
---|---|
Molecular Formula | C8H10 |
MDL Number | MFCD00077264 |
Synonym | Dimethylbenzene |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC200910010
|
Thermo Scientific Chemicals
200910010 |
1 L | Glass Bottle |
Each for $128.97
Case of 4 Each for $0.00
|
|
|||||
AC200910025
|
Thermo Scientific Chemicals
200910025 |
2.5 L | Glass Bottle |
Each for $203.72
Case of 4 Each for $0.00
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.
Chemical Identifiers
1330-20-7 | |
C8H10 | |
MFCD00077264 | |
Dimethylbenzene |
Specifications
-34.0°C | |
0.8650g/mL | |
1 L | |
Authentic | |
Liquid | |
96% | |
C8H10 | |
MFCD00077264 | |
0.6 mPa.s (20°C) | |
15, 10278 | |
106.17 | |
Pure |
1330-20-7,100-41-4 | |
136.0°C to 140.0°C | |
23°C | |
Glass Bottle | |
1.4940 to 1.4980 (20°C, 589nm) | |
0.865 | |
C6H4(CH3)2 | |
Vapor Pressure: 8mbar at 20°C | |
05, 360 | |
Dimethylbenzene | |
96% | |
Xylenes, Mixed Isomers with Ethylbenzene |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 215-535-7
RTECSNumber : ZE2100000
TSCA : TSCA
RUO – Research Use Only