missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylenes, ACS, 98.5+% (Assay, isomers plus ethylbenzene), Thermo Scientific Chemicals

$60.05 - $213.30

Chemical Identifiers

CAS 1330-20-7
Molecular Formula (CH3)2C6H4
MDL Number MFCD00077264
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA16371AP
View Documents
Thermo Scientific Chemicals
016371AP
500 mL
Each for $60.05
 
AA16371K2
View Documents
Thermo Scientific Chemicals
016371K2
1 L
Each for $76.63
 
AA16371M1
View Documents
Thermo Scientific Chemicals
016371M1
4 x 1 L
Each for $213.30
 
AA16371K7
View Documents
Thermo Scientific Chemicals
016371K7
4 L
Each for $188.32
 
Description

Description

Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1330-20-7
(CH3)2C6H4
MFCD00077264

Specifications

<°C to 25°C
Colorless
0.86 g/cm3
500 mL
465°C (869°F)
1.497
(CH3)2C6H4
1901563
106.17
Xylenes, For ACS analysis
1330-20-7
Colorless liquid
137°C to 144°C
29°C (84°F)
Liquid
>98.5%
MFCD00077264
Miscible with aromatic hydrocarbons. Immiscible with water.
ACS
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H312-H332-H315;
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H226-H304-H312+H332-H315-H319-H335-H373

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES

EINECSNumber : 215-535-7

RTECSNumber : ZE2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only