missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Xylenes, Reagent ACS, 99%, Thermo Scientific Chemicals

$91.95 - $965.42

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Dimethylbenzene
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC422685000
SDS View Documents
Thermo Scientific Chemicals
422685000
500 mL
Each for $91.95
Case of 6 Each for $0.00
 
AC422680025
SDS View Documents
Thermo Scientific Chemicals
422680025
2.5 L
Each for $211.41
 
AC422680040
SDS View Documents
Thermo Scientific Chemicals
422680040
4 L
Each for $302.75
 
AC422680200
SDS View Documents
Thermo Scientific Chemicals
422680200
20 L
Each for $965.42
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Specifications

Chemical Identifiers

1330-20-7
C8H10
MFCD00077264
Dimethylbenzene

Specifications

-34.0°C
0.8650g/mL
500 mL
Authentic
Liquid
0.002% max.
0.865
C6H4(CH3)2
Vapor Pressure: 8mbar at 20°C
0.05% max. (Coulometric)
15, 10278
Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride, acetone and ether
99%
Xylenes
1330-20-7,100-41-4
136.0°C to 140.0°C
23°C
Glass Bottle
1.4940 to 1.4980
99%
C8H10
MFCD00077264
0.6 mPa.s (20°C)
05, 360
Dimethylbenzene
106.17
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-535-7

RTECSNumber : ZE2100000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only