missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenol Orange Disodium Salt, For spectrophotometric determination of metal ions, MilliporeSigma™ Supelco™
$102.33 - $385.29
Chemical Identifiers
| CAS | 1611-35-4 |
|---|---|
| Molecular Formula | C31H32N2O13S |
| Molecular Weight (g/mol) | 672.66 |
| InChI Key | ORZHVTYKPFFVMG-UHFFFAOYSA-N |
| Synonym | 3,3 '-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt |
| IUPAC Name | 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λâ¶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid |
| SMILES | CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1 |
Chemical Identifiers
| 1611-35-4 | |
| 672.66 | |
| 3,3 '-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt | |
| CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1 |
| C31H32N2O13S | |
| ORZHVTYKPFFVMG-UHFFFAOYSA-N | |
| 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λâ¶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid |
Specifications
| 1611-35-4 | |
| C31H30N2Na2O13S | |
| ORZHVTYKPFFVMG-UHFFFAOYSA-N | |
| 2-[({5-[3-(3-{[bis(carboxymethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-1,1-dioxo-3H-2,1λâ¶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxymethyl)amino]acetic acid | |
| 716.62 |
| C31H32N2O13S | |
| 3,3 '-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein disodium salt | |
| CC1=CC(=CC(CN(CC(O)=O)CC(O)=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(CN(CC(O)=O)CC(O)=O)=C1 | |
| 672.66 | |
| 1 g |