missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Xylometazoline hydrochloride, 98%
Chemical Identifiers
CAS | 1218-35-5 |
---|---|
Molecular Formula | C16H24N2·HCL |
Molecular Weight (g/mol) | 280.84 |
InChI Key | YGWFCQYETHJKNX-UHFFFAOYSA-N |
Synonym | xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride |
PubChem CID | 5282386 |
IUPAC Name | 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride |
SMILES | CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC461990050
|
Thermo Scientific™
461990050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC461990250
|
Thermo Scientific™
461990250 |
25g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1218-35-5 | |
280.84 | |
xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride | |
2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride |
C16H24N2·HCL | |
YGWFCQYETHJKNX-UHFFFAOYSA-N | |
5282386 | |
CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl |
Specifications
1218-35-5 | |
5g | |
(5% in water) Clear colorless to light yellow | |
CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl | |
280.84 | |
280.84 | |
Xylometazoline hydrochloride |
C16H24N2·HCL | |
xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride | |
YGWFCQYETHJKNX-UHFFFAOYSA-N | |
2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride | |
5282386 | |
98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 214-936-4
RUO â Research Use Only