missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Xylose, USP, 98-102%, Spectrum™ Chemical
XY110, 58-86-6, C5H10O5
Supplier: Spectrum Chemical Mfg Cor XY11025KGBL
Description
Spectrum™ Chemical Xylose, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
58-86-6 | |
0.0005 | |
Fiber Drum | |
25 kg | |
SRBFZHDQGSBBOR-UHFFFAOYNA-N | |
oxane-2,3,4,5-tetrol | |
98 to 102% |
1 | |
0.001 | |
C5H10O5 | |
+18.2° to +19.4° | |
OC1COC(O)C(O)C1O | |
150.13 | |
USP |
Chemical Identifiers
58-86-6 | |
150.13 | |
oxane-2,3,4,5-tetrol |
C5H10O5 | |
SRBFZHDQGSBBOR-UHFFFAOYNA-N | |
OC1COC(O)C(O)C1O |