Youngstown Barrel & Drum Fiber Drums
Youngstown Barrel & Drum fiber drums are constructed of sturdy fiberboard and are an excellent solution for the containment and storage of dry or solid materials.
Supplier:  Youngstown Barrel & Drum YBD2013
 View more versions of this product
Catalog No. 18120246
Explore available promotions
Description
Description
  • Open-head drums qualify for transport of dry, nonreactive solid materials only
  • Available with all-fiber construction (fiber covers) or with steel chime bands and metal cover
Specifications
Specifications
Fiber Drum
45 L
Fiber
22.62 in.
22.62 in.
4.5 lb.
UN 1G/X60/S
For transport of dry, nonreactive solid materials only
Open
57.45 cm
57.45 cm
2.04 Kg
Product Suggestions
Product Suggestions