YSI™ DO Cap Membrane Kit
DO Cap Membrane Kit for use with probes and electrodes.
$207.24 - $264.94
Specifications
Type DO Cap Membrane Kit
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Includes Material Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Includes Material Price Quantity  
15-176-212
View Documents
YSI™
605914
Probes: Pro Series Galvanic Sensors Newer Membrane Blue Cap Membrane, 2.0 mil Polyethylene
Each for $207.24
15-177-560
View Documents
YSI™
059880
Instrument: 85, 550, and 556 MPS. Probes: 5239, 5905, 5010 and Pro Series Polarographic Sensors Black Cap Membrane, 1.0 mil PTFE
Pack of 6 Each for $264.94
Description
Description
  • DO Cap Membrane Kit
  • For use with sensors and probes
Specifications
Specifications
DO Cap Membrane Kit
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'