YSI™ Membrane kits
$202.72 - $218.15
Catalog Number | Mfr. No. | Probe Type | Includes | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Probe Type | Includes | Price | Quantity | ||||
14-660-104
|
YSI™
5909 |
Oxygen/Temperature | Blue 2 mil. thickness membranes, 6 cap membranes and KCl solution |
Pack of 6 Each for $218.15
|
|
||||
14-660-103
|
YSI™
YSI605306 |
DO | Six 1.25 mil. cap membranes, KCl solution |
Pack of 6 Each for $202.72
|
|
||||
Specifications
YSI™ Model 556 | |
Membrane Kit |