YSI™ Pro Plus Quatro Field Cable
For use with Professional Plus instrument.
Supplier: YSI™ 60579030
Includes: Conductivity/temperature sensor
Description
- 4-port cable
- True field cable with MS (military spec) connectors
- Rugged, weighted probe guard
- Measure conductivity/temperature, dissolved oxygen and any two ISEs among pH, ORP, ammonium, nitrate or chloride
- Cable Length: 30m
Warranty
Two years
Specifications
Professional Plus instrument |