YSI™ Pro Series DO Cap Membrane Kit
Supplier:  YSI™ YSI605913
 View more versions of this product
Catalog No. 15-176-211
Explore available promotions
Description
Description
Specifications
Specifications
Membrane Kit
For pro series galvanic sensors
Bottle of NaCl electrolyte, 1 sanding disc, six screw-on cap 1.25mL Polyethylene make DO membranes, yellow color