YSI™ ProODO Optical DO Cable
For use with ProODO, ProDSS, and 5500D instruments.
Supplier:  YSI™ 626250100
Catalog No. 14-660-214
Explore available promotions
Description
Description
  • ODO Digital probe assembly with cable
  • Includes ODO optical DO sensor, sensing cap, cable management kit
  • For ODO and temperature
  • Cable Length: 100m
Specifications
Specifications
ODO optical DO sensor, sensing cap, cable management kit
Optical DO Cable