missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

YSI™ TruLab™ pH/ISE 1320 Dual Channel Laboratory ISE Benchtop Meter

Easy-to-use meter for dual measurement of pH, mV (ORP) or ISE and temperature

Supplier:  YSI™ YSI1GA340Y

Catalog No. 15-177-829


Explore available promotions

Includes: Meter, universal power supply, electrode stand, manual, PC software and USB cable

Description

Description

  • Provides automatic documentation for GLP compliance
  • AutoRead function improves repeatability
  • Easy-to-read backlit display with text menu for easy operation for pH, mV (ORP), ISE and temperature
  • Fast data transfer via USB in .csv or ASCII format or the optional integrated printer
  • Easy calibration with adjustable calibration timer which indicates when to perform the next calibration routine
  • Automatic or manual temperature compensation
  • 1, 2, 3, 4 or 5 point calibration for pH
  • 1, 2, 3, 4, 5, 6 or 7 point calibration for ISE, also non- linear
  • BNC connection for pH, mV and ISEs; banana connection for temperature probes (sold separately)
  • Continuous Monitoring Control (CMC) function for monitoring the ideal measuring range for pH measurements based on the last calibration
  • Wide selection of electrodes: TruLab analog pH/mV/ISE/temp probes
  • Four alkaline AA batteries provide approx.150 hours with illumination; universal power supply; also compatible with four NiMH AA rechargeables
Parameters Measured:
  • pH, mV (ORP/Redox)
  • Temperature
  • ISEs: Ammonia, Ammonium, Bromide, Cadmium, Calcium, Chloride, Copper, Cyanide, Fluoride, Iodide, Lead, Nitrate, Potassium, Silver/Sulfide, Sodium
Specifications:
  • Buffer Recognition: Yes
  • Sampling: Yes
  • User Calibratable: Yes
  • Logging Capabilities: Yes
  • pH Range: -2.0 to 20.0; -2.00 to 20.00; -2.000 to 19.999
  • pH Resolutions: 0.1; 0.01; 0.001
  • pH Accuracy (±1 digit): ±0.1; ±0.01; ±0.005 (temp. of test sample 15° to 35°C; within a range of ±2 pH of the calibration point)
  • mV Range: -1200 to 1200.0; -2500 to 2500
  • mV Resolution: 0.1; 1.0
  • mV Accuracy: ±0.3; ±1.0 (temp. of test sample 15° to 35°C)
  • ISE Range (mg/L, mg/kg, mmol/L, ppm, %): 0.000 to 9.999; 10.00 to 99.99; 100.0 to 999.9; 1000 to 999999
  • ISE Range (μmol/L): 0.000 to 9.999; 10.00 to 99.99; 100.0 to 999.9; 1000 to 9999
  • ISE Resolution (mg/L, mg/kg, mmol/L, ppm, %): 0.001; 0.01; 0.1; 1
  • ISE Resolution (μmol/L): 0.001; 0.01; 0.1; 1
  • ISE Accuracy: ±0.3; ±1.0 (temp of test sample 15° to 35°C)
  • Temperature Range: -5° to +105°C (23° to 221°F)
  • Temperature Resolution: 0.1
  • Temperature Accuracy: ±0.1
  • Memory: 5000 data sets auto logging, 500 data sets manual logging
  • Manual Temperature Input: -25° to +130°C (-13° to +266°F) in increments of 1
  • Storage Temperature: -25° to +65°C (-13° to +149°F)
  • Unit of Measure: Sample subtraction, Known addition (double), Sample addition, ISE Measurements: Blank value correction, increment methods: Known addition (single), Known subtraction
  • Weight: 0.8kg (1.8 lb.)
  • Dimensions (L x W x H): 23 x 19 x 8cm (9 x 7.5 x 3.2 in.)

Order Info

Cables, probes/sensors, and accessories sold separately.

Certifications

IP43, electrical protective class III, cETLus, CE

Compliance

GLP/AQA compliant

Warranty

Three years

Specifications

Specifications

±0.3; ±1.0 (temp. of test sample 15° to 35°C)
±0.1
4 x alkaline AA batteries (also compatible with four NiMH AA rechargeables)
CE, cETLus; electrical protective class III; IP43
2 to 7
-1200 to 1200.0, -2500 to 2500
0.001, 0.01, 0.1, 1 μmol/L, 0.001, 0.01, 0.1, 1 in mg/L, mg/kg, mmol/L, ppm, %
-5°C to +105°C
Measure pH, mV (ORP/Redox); Temperature; ISEs (Ammonia, Ammonium, Bromide, Cadmium, Calcium, Chloride, Copper, Cyanide, Fluoride, Iodide, Lead, Nitrate, Potassium, Silver/Sulfide, Sodium)
Dual channel
0.1; 1.0
pH/ISE 1320 Laboratory ISE Benchtop Meter
Universal power supply
±0.1; ±0.01; ±0.005 (temp. of test sample 15° to 35°C; within a range o
approx.150 hours with illumination, 1000 hours without
1, 2, 3, 4 or 5 point with or without autobuffer recognition; USA and NIST and 22 pre-programmed buf
5000 data sets auto logging, 500 data sets manual logging
0.000 to 9.999, 10.00 to 99.99, 100.0 to 999.9, 1000 to 999999 μmol/L, 0.000 to 9.999, 10.00 to 99.99, 100.0 to 999.9, 1000 to 9999 in mg/L, mg/kg, mmol/L, ppm, %
-2.0 to 20.0, -2.00 to 20.00, -2.000 to 19.999
0.1, 0.01, 0.001
Color Graphic Display
Meter, universal power supply, electrode stand, manual, PC software and USB cable
Four alkaline AA batteries fully charged provides approx.150 hours of illumination; universal power supply; also compatible with four NiMH AA rechargeables
0.1
3 Years
Videos
SDS
Documents

Documents

Promotions

Promotions