Learn More
Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO), Thermo Scientific Chemicals
$199.12 - $712.67
Chemical Identifiers
CAS | 7440-64-4 |
---|---|
Molecular Formula | Yb |
Molecular Weight (g/mol) | 173.05 |
MDL Number | MFCD00011286 |
InChI Key | NAWDYIZEMPQZHO-UHFFFAOYSA-N |
Synonym | unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n |
PubChem CID | 23992 |
ChEBI | CHEBI:33381 |
IUPAC Name | ytterbium |
SMILES | [Yb] |
Description
Used for phosphors, ceramic capacitors, ferrite devices, pulsed lasers and catalysts. Use in improving the strength and mechanical properties of stainless steel. Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7440-64-4 | |
173.05 | |
NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
23992 | |
ytterbium |
Yb | |
MFCD00011286 | |
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
CHEBI:33381 | |
[Yb] |
Specifications
7440-64-4 | |
MFCD00011286 | |
25 x 25 mm | |
It reacts slowly with water and is soluble in dilute acids. | |
[Yb] | |
173.05 | |
CHEBI:33381 | |
Odorless | |
≈0.43g/25x25mm | |
Ytterbium foil, 0.1mm (0.004 in.) thick |
Yb | |
UN3178 | |
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n | |
NAWDYIZEMPQZHO-UHFFFAOYSA-N | |
ytterbium | |
23992 | |
99.9% | |
Mylar under Argon | |
Foil | |
(REO) |
Safety and Handling
GHS H Statement
H260-H228
In contact with water releases flammable gases which may ignite spontaneously.
Flammable solid.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber : 231-173-2
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only