missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ytterbium foil, 1.0mm (0.04in) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 010426FF

 View more versions of this product

Catalog No. AA10426FF


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Ytterbium foil
7440-64-4
Yb
25 x 25 mm
NAWDYIZEMPQZHO-UHFFFAOYSA-N
ytterbium
23992
99.9%
Mylar under Argon
Foil
1.0mm (0.04 in.) thick
(REO)
MFCD00011286
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
[Yb]
173.05
CHEBI:33381
Odorless
≈4.4g/25x25mm

Chemical Identifiers

7440-64-4
173.05
NAWDYIZEMPQZHO-UHFFFAOYSA-N
23992
ytterbium
Yb
MFCD00011286
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
CHEBI:33381
[Yb]
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 231-173-2

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only